missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 4.5, Ricca Chemical

Catalog No. R005500020F
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R005500020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R005500020F Supplier Ricca Chemical Company Supplier No. R005500020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use pH 4.5 acetate buffer for consistent pH control in laboratory applications.

Specifications

Chemical Name or Material Acetate Buffer
CAS 7732-18-5, 6131-90-4, 64-19-7
Color Colorless Liquid
CAS Max % 99.53, 0.3, 0.17
Concentration 1X
For Use With (Application) Water Analysis
pH 4.5
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Sterility Non-sterile
Grade Laboratory
Show More Show Less