missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer pH 4.5, For Cyanide, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Acetate Buffer pH 4.5 |
| Name Note | For Cyanide |
| CAS | 64-19-7, 6131-90-4, 7732-18-5 |
| Color | Colorless |
| CAS Max % | 36.9, 33.6, 29.5 |
| Decomposition Information | Carbon monoxide, Carbon dioxide |
| pH | 4.50 ±0.05 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Identification | Passes Test |
| Show More |