missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 4.0 for Residual Chlorine Analysis, Reagents

Catalog No. 01336010
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336010 1 L
01336011 10 L
01336009 4 L
01336008 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336010 Supplier Labchem Inc Supplier No. CA0164001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetate Buffer, pH 4.0 for Residual Chlorine Analysis, Reagent, use for Water and wastewater analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Acetate Buffer, pH 4.0 for Residual Chlorine Analysis
CAS 64-19-7, 7732-18-5, 7732-18-5
Boiling Point 118°C
Color Colorless
pH 4
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.15
Sterility Non-sterile
Grade Reagent
Show More Show Less