missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer pH 4.0, For Chlorine, Iodine, Certified, LabChem™

Catalog No. LC100302
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC100302 1 L
LC100301 500 mL
LC100304 4 L
LC100306 10 L
LC100305 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC100302 Supplier LabChem Supplier No. LC100302
Only null left

Specifications

Chemical Name or Material Acetate Buffer pH 4.0
Name Note For Chlorine, Iodine
CAS 64-19-7, 7732-18-5, 6131-90-4
Color Colorless
CAS Max % 45.5, 32.5, 22
Decomposition Information Carbon monoxide, Carbon dioxide
pH 4.0 ±0.1
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula CH3COOHNa·3H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.08
Grade Certified
Show More Show Less