missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer pH 4.0, For Chlorine, Iodine, Certified, LabChem™

Catalog No. LC100301
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC100301 500 mL
LC100302 1 L
LC100304 4 L
LC100306 10 L
LC100305 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC100301 Supplier LabChem Supplier No. LC100301
Only null left

Specifications

Chemical Name or Material Acetate Buffer pH 4.0
Name Note For Chlorine, Iodine
CAS 64-19-7, 7732-18-5, 6131-90-4
Color Colorless
CAS Max % 45.5, 32.5, 22
Decomposition Information Carbon monoxide, Carbon dioxide
pH 4.0 ±0.1
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Linear Formula CH3COOHNa·3H2O
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 136.08
Grade Certified
Show More Show Less