missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, for Iron (Phenanthroline Method), Ricca Chemical

Catalog No. 6032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6032 1 L Natural Poly Bottle
6016 500 mL Natural Poly Bottle
601 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6032 Supplier Ricca Chemical Company Supplier No. RI6032
Only null left

For Analysis of Iron (pHenanthroline Method)

Specifications

Chemical Name or Material Acetate Buffer
CAS 64-19-7, 631-61-8, 7732-18-5
CAS Min % 63.0, 21.6, 12.95
Color Colorless
CAS Max % 66.0, 22.48, 13.48
Concentration 1X
For Use With (Application) Iron Analysis
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less