missing translation for 'onlineSavingsMsg'
Learn More

Acetanilide, 99%, Spectrum™ Chemical

Catalog No. 18608318
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608318 250 g Poly Pail
18608320 500 g Poly Pail
18608316 1 kg Poly Pail
18608317 12 kg Poly Pail
18608319 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608318 Supplier Spectrum Chemical Mfg Cor Supplier No. A1007250GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 103-84-4

Specifications

CAS Max % 1
Melting Point 113° to 118°C
Boiling Point 304°C
Packaging Poly Pail
Quantity 250 g
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.