missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetanilide, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor A100712KGBL
Specifications
| 103-84-4 | |
| 113° to 118°C | |
| Poly Pail | |
| 0.99 | |
| Crystalline Powder or Lumps |
| 1 | |
| 304°C | |
| 12 kg | |
| Reagent |
Chemical Identifiers
| 103-84-4 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.