missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Acetaminophen, USP
Description
Acetaminophen USP is a white, odorless, crystalline powder.
Alternative name(s): paracetamol
Specifications
Specifications
| Chemical Name or Material | Acetaminophen, USP |
| Packaging | Pail, 8 Liter, White, HDPE, Square |
| CAS | 103-90-2 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 151.1641 |
| Quantity | 2.5 kg |
| Percent Purity | 98 to 102 % |
| Grade | USP |