missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. p-7187388
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012355 1 g
111002436 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012355 Supplier Merck Emd Millipore Supplier No. PHR10051G
Only null left

Acetaminophen is a drug, which possesses pharmacological properties like antipyretic and analgesic property.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 103-90-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3 CONHC6H4 OH
MDL Number MFCD00002328
Melting Point 168°C to 172°C
Synonym 4′-Hydroxyacetanilide; 4-Acetamidophenol; N-(4-Hydroxyphenyl)acetamide; N-Acetyl-4-aminophenol; APAP; Paracetamol
Type Certified Reference Material
Quantity 1 g
Formula Weight 151.16
Grade Certified Reference Material
Show More Show Less