missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Impurity I, Pharmaceutical Secondary Standards, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012708
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012708 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012708 Supplier Merck Emd Millipore Supplier No. PHR1973100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 118-93-4
Vapor Pressure ˜0.2 mmHg (20°C)
Vapor Density 4.7 (vs. air)
Density 1.131 g/mL (at 25°C)
Linear Formula HOC6H4COCH3
MDL Number MFCD00002219
Melting Point 3°C to 6°C
Boiling Point 213°C (717 mmHg)
Synonym 2 '-Hydroxyacetophenone; 2 '-Hydroxyacetophenone; 2-Acetylphenol
Quantity 100 mg
Formula Weight 136.15
Grade Certified Reference Material
Show More Show Less