missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen Impurity G, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111003817
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003817 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003817 Supplier Merck Emd Millipore Supplier No. PHR1972100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

Linear Formula C8H9NO2
Synonym 1-(4-hydroxyphenyl)ethanone oxime; 1-(4-hydroxyphenyl)ethanone oxime
Quantity 100 mg
Formula Weight 151.16
Grade Certified Reference Material