missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaminophen Impurity E Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 99-93-4 |
| Linear Formula | HOC6H4 COCH3 |
| MDL Number | MFCD00002359 |
| Melting Point | 109°C to 111°C |
| Boiling Point | 147°C to 148°C (3 mmHg) |
| Synonym | 4 ′-Hydroxyacetophenone; 4 ′-Hydroxyacetophenone; 4-Hydroxyphenylethanone; p-Acetophenol; p-Hydroxyphenyl methyl ketone; Piceol |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 136.15 |
| Grade | Certified Reference Material |