missing translation for 'onlineSavingsMsg'
Learn More

Acetamidoxime 97.0+%, TCI America™

Catalog No. p-7118004
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26465G 5 g
A264625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26465G Supplier TCI America Supplier No. A26465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22059-22-9
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00603514 MFCD00603514
InChI Key AEXITZJSLGALNH-UHFFFAOYSA-N
Synonym n'-hydroxyacetimidamide, n-hydroxyacetamidine, methylamidoxime, acetamide, oxime, 1z-n'-hydroxyethanimidamide, n-hydroxyethanimidamide, oximinoacetamide, e-n'-hydroxyacetimidamide, n'-hydroxyacetamidine, n-hydroxy-acetamidine
PubChem CID 5487681
IUPAC Name (Z)-N'-hydroxyethanimidamide
SMILES C\C(N)=N\O

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Formula Weight 74.08
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetamidoxime
RTECSNumber AC6900000
TSCA No