missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetamidine Hydroiodide (Low water content) 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 186.00 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Acetamidine Hydroiodide (Low water content) |
Safety and Handling
| TSCA | No |