accessibility menu, dialog, popup

UserName

CH3NHH2NHI

CAS RN 1452099-14-7

IUPAC Name: ethanimidamide hydroiodide
Synonyms: acetamidine hydroiodide
Molecular Weight (g/mol): 186
Molecular Formula: C2H7IN2
InChi Key: GGYGJCFIYJVWIP-UHFFFAOYSA-N
SMILES: I.CC(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results