missing translation for 'onlineSavingsMsg'
Learn More

Acetamidine Hydroiodide (Low water content) 98.0+%, TCI America™

Catalog No. A29021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29021G 1 g
A29025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29021G Supplier TCI America Supplier No. A29021G
Only null left

Chemical Identifiers

CAS 1452099-14-7
Molecular Formula C2H7IN2
Molecular Weight (g/mol) 185.996
InChI Key GGYGJCFIYJVWIP-UHFFFAOYSA-N
Synonym Acetamidinium Iodide
PubChem CID 91972120
IUPAC Name ethanimidamide;hydroiodide
SMILES CC(=N)N.I

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 186.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetamidine Hydroiodide (Low water content)