missing translation for 'onlineSavingsMsg'
Learn More

Acetamide oxime, 95+%

Catalog No. p-3559311
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC390530010 1 g Glass bottle
AC390530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC390530010 Supplier Thermo Scientific Chemicals Supplier No. 390530010
Only null left

CAS: 22059-22-9 | C2H6N2O | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22059-22-9
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00603514 MFCD00603514
InChI Key AEXITZJSLGALNH-UHFFFAOYSA-N
Synonym n'-hydroxyacetimidamide, n-hydroxyacetamidine, methylamidoxime, acetamide, oxime, 1z-n'-hydroxyethanimidamide, n-hydroxyethanimidamide, oximinoacetamide, e-n'-hydroxyacetimidamide, n'-hydroxyacetamidine, n-hydroxy-acetamidine
PubChem CID 5487681
IUPAC Name N'-hydroxyethanimidamide
SMILES C\C(N)=N\O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 134.0°C to 138.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Linear Formula CH3C(NH2)=NOH
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,insoluble in ether and pentane
Formula Weight 74.08
Percent Purity 95+%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Acetamide oxime
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only