missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime 99.0+%, TCI America™

Catalog No. A000625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A000625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A000625ML Supplier TCI America Supplier No. A000625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (E)-N-ethylidenehydroxylamine
SMILES C\C=N\O

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 2332
Formula Weight 59.07
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetaldoxime
RTECSNumber AB2975000
TSCA Yes