accessibility menu, dialog, popup

UserName

CAS RN 107-29-9

IUPAC Name: (E)-N-ethylidenehydroxylamine
Synonyms: acetaldoxime
Molecular Weight (g/mol): 59.07
Molecular Formula: C2H5NO
InChi Key: FZENGILVLUJGJX-NSCUHMNNSA-N
SMILES: C\C=N\O
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)