missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, syn + anti, 98%

Catalog No. p-7080906
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1064018 Supplier Thermo Scientific Chemicals Supplier No. A1064018
Only null left

CAS: 107-29-9 | C2H5NO | 59.07 g/mol

Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

Solubility
Soluble in water, ethanol and ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (NZ)-N-ethylidenehydroxylamine
SMILES C\C=N\O

Specifications

Density 0.969
Boiling Point 115°C to 117°C
Flash Point 36°C (96°F)
Odor Characteristic
Refractive Index 1.426
Quantity 50 g
UN Number UN2332
Beilstein 1209252
Merck Index 14,42
Solubility Information Soluble in water,ethanol and ether.
Formula Weight 59.07
Percent Purity 98%
Chemical Name or Material Acetaldoxime, syn + anti
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H226-H302-H312-H332-H319
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ACETALDEHYDE OXIME
EINECSNumber 203-479-6
RTECSNumber AB2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only