missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, 99%, mixture of syn and anti

CAS: 107-29-9 | C2H5NO | 59.07 g/mol

$254.11 - $254.11

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (NZ)-N-ethylidenehydroxylamine
SMILES C\C=N\O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102231000
SDS View Documents
Thermo Scientific Chemicals
102231000
100 g Glass bottle
Each for $254.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-29-9
59.07
FZENGILVLUJGJX-NSCUHMNNSA-N
5324280
(NZ)-N-ethylidenehydroxylamine
C2H5NO
MFCD00002124 MFCD00002124
acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
CHEBI:50719
C\C=N\O

Specifications

107-29-9
100.0
White
115.0°C
Authentic
Glass bottle
1.4250 to 1.4270
MFCD00002124 MFCD00002124
01, IV, 3121
0.96
acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
FZENGILVLUJGJX-NSCUHMNNSA-N
(NZ)-N-ethylidenehydroxylamine
5324280
59.06
Liquid or Low Melting Solid
98.5
44.0°C to 46.0°C
0.9600g/mL
38°C
98.5% min. (GC)
C2H5NO
CH3CH=NOH
100 g
01,3
15, 43
Solubility in water: soluble in water. Other solubilities: decomposes by aq. hcl,soluble in alcohol and ether
C\C=N\O
59.07
CHEBI:50719
99%
Acetaldoxime, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-479-6

RTECSNumber : AB2975000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only