missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC S125
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Acetaldehyde | 75-07-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Acetaldehyde | |
| 75-07-0,67-56-1 | |
| 0.1,99.9 | |
| Acetaldehyde | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| IKHGUXGNUITLKF-UHFFFAOYSA-N | |
| CC=O | |
| acetaldehyde | |
| 1 mL | |
| CHEBI:15343 |
| 2mL Amber Prescored Glass Ampule | |
| Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. | |
| 0.1,99.9 | |
| C2H4O | |
| Certified Reference Material | |
| Acetaldehyde | |
| Methanol-P and T | |
| 44.053 | |
| 177 |
Chemical Identifiers
| 75-07-0 | |
| 44.053 | |
| 177 | |
| acetaldehyde |
| C2H4O | |
| IKHGUXGNUITLKF-UHFFFAOYSA-N | |
| CHEBI:15343 | |
| CC=O |
Safety and Handling
Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only