missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde Semicarbazone 98.0+%, TCI America™
$90.48 - $269.52
Chemical Identifiers
| CAS | 591-86-6 |
|---|---|
| Molecular Formula | C3H7N3O |
| Molecular Weight (g/mol) | 101.109 |
| MDL Number | MFCD00059191 |
| InChI Key | IUNNCDSJWDQYPW-GORDUTHDSA-N |
| PubChem CID | 5381348 |
| IUPAC Name | [(E)-ethylideneamino]urea |
| SMILES | CC=NNC(=O)N |
Chemical Identifiers
| 591-86-6 | |
| 101.109 | |
| IUNNCDSJWDQYPW-GORDUTHDSA-N | |
| [(E)-ethylideneamino]urea |
| C3H7N3O | |
| MFCD00059191 | |
| 5381348 | |
| CC=NNC(=O)N |
Specifications
| 591-86-6 | |
| White | |
| MFCD00059191 | |
| IUNNCDSJWDQYPW-GORDUTHDSA-N | |
| [(E)-ethylideneamino]urea | |
| 5381348 | |
| ≥98.0% (T) | |
| Acetaldehyde Semicarbazone |
| 164°C | |
| C3H7N3O | |
| 5 g | |
| CC=NNC(=O)N | |
| 101.109 | |
| 101.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No