accessibility menu, dialog, popup

UserName

CH3NNHH2NO(Z)

CAS RN 591-86-6

IUPAC Name: [(Z)-ethylideneamino]urea
Synonyms: (Z)-ethylideneaminourea
Molecular Weight (g/mol): 101.11
Molecular Formula: C3H7N3O
InChi Key: IUNNCDSJWDQYPW-DJWKRKHSSA-N
SMILES: C\C=N/NC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results