missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame K Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014602
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014602 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014602 Supplier Merck Emd Millipore Supplier No. PHR1266500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acesulfame K is a synthetically prepared sulfonyl amide sweetener, widely used with sugar alcohols in food and beverage industry.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 55589-62-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C4H4 KNO4 S
MDL Number MFCD00043833
Synonym 6-Methyl-1,2,3-oxathiazin-4(3 H)-one 2,2-dioxide potassium salt
Type Certified Reference Material
Quantity 500 mg
Formula Weight 201.24
Grade Certified Reference Material