missing translation for 'onlineSavingsMsg'
Learn More

Acesulfame K for food analysis, 99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014600
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014600 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014600 Supplier Merck Emd Millipore Supplier No. 0405425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages

Specifications

CAS 55589-62-3
Linear Formula C4H4 KNO4 S
MDL Number MFCD00043833
Synonym 6-Methyl-1,2,3-oxathiazin-4(3 H)-one 2,2-dioxide potassium salt
Type Food Analysis Standard
Quantity 25 g
Formula Weight 201.24
Percent Purity ≥99% (HPLC)
Grade Food Analysis Standards