missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthylene 94.0+%, TCI America™

Catalog No. p-7116652
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00055G 5 g
A000525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A00055G Supplier TCI America Supplier No. A00055G
Only null left

Chemical Identifiers

CAS 208-96-8
Molecular Formula C12H8
Molecular Weight (g/mol) 152.20
MDL Number MFCD00003806
InChI Key HXGDTGSAIMULJN-UHFFFAOYSA-N
Synonym acenaphthalene, cyclopenta de naphthalene, dsstox_cid_3845, dsstox_rid_77204, dsstox_gsid_23845, acenaphthylen, acenaphthylene, radical ion 1-, acenaphthylene
PubChem CID 9161
ChEBI CHEBI:33081
IUPAC Name acenaphthylene
SMILES C1=CC2=C3C1=CC=CC3=CC=C2

Specifications

Melting Point 93°C
Boiling Point 275°C
Quantity 5 g
Formula Weight 152.20
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Acenaphthylene
EINECSNumber (4)-0644
RTECSNumber AB1254000
TSCA Yes