missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acenaphthylene 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Boiling Point | 275°C |
| Quantity | 25 g |
| Formula Weight | 152.20 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Acenaphthylene |
Safety and Handling
| EINECSNumber | (4)-0644 |
| RTECSNumber | AB1254000 |
| TSCA | Yes |