missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone Dioxime, TCI America™

Catalog No. A09815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A09815G Supplier TCI America Supplier No. A09815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8-7-1932
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.21
MDL Number MFCD00059513
InChI Key DFVXPXWNTZFHIV-UHFFFAOYSA-N
PubChem CID 233285
IUPAC Name N-(2-nitrosoacenaphthylen-1-yl)hydroxylamine
SMILES C1=CC2=C3C(=C1)C(=C(C3=CC=C2)N=O)NO

Specifications

Quantity 5 g
Formula Weight 212.21
Physical Form Crystalline Powder
Chemical Name or Material Acenaphthenequinone Dioxime