missing translation for 'onlineSavingsMsg'
Learn More

Acemetacin

Catalog No. p-7042737
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6358304 2 g
AAJ6358309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6358304 Supplier Thermo Scientific Chemicals Supplier No. J6358304
Only null left

A potent inhibitor of COX-2 found to have anti-tumor activity in the colon | CAS: 53164-05-9 | C21H18ClNO6 | 415.826 g/mol

A potent inhibitor of COX-2 found to have anti-tumor activity in the colon

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53164-05-9
Molecular Formula C21H18ClNO6
Molecular Weight (g/mol) 415.826
MDL Number MFCD00151473
InChI Key FSQKKOOTNAMONP-UHFFFAOYSA-N
Synonym acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester
PubChem CID 1981
ChEBI CHEBI:31162
IUPAC Name 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O

Specifications

Melting Point 149°C to 153°C
Color Yellow
Quantity 2 g
UN Number UN2811
Merck Index 14,27
Formula Weight 415.83
Physical Form Powder
Chemical Name or Material Acemetacin
Hazard Category H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 258-403-4
RTECSNumber NL3521400
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only