missing translation for 'onlineSavingsMsg'
Learn More

Acemetacin 98.0+%, TCI America™

Catalog No. p-7117918
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24525G 5 g
A245225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24525G Supplier TCI America Supplier No. A24525G
Only null left

Chemical Identifiers

CAS 53164-05-9
Molecular Formula C21H18ClNO6
Molecular Weight (g/mol) 415.826
MDL Number MFCD00151473
InChI Key FSQKKOOTNAMONP-UHFFFAOYSA-N
Synonym acemetacin, rantudil, emflex, acemetacinum, acemix, acemetacina, acemetacine, aximeixin, rheumibis, indomethacin carboxymethyl ester
PubChem CID 1981
ChEBI CHEBI:31162
IUPAC Name 2-[2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetyl]oxyacetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC(=O)O

Specifications

Melting Point 152°C
Quantity 5 g
UN Number 2811
Formula Weight 415.83
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acemetacin
RTECSNumber NL3521400
TSCA No