missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aceclofenac 98.0+%, TCI America™
$37.29 - $82.02
Chemical Identifiers
| CAS | 89796-99-6 |
|---|---|
| Molecular Formula | C16H13Cl2NO4 |
| Molecular Weight (g/mol) | 354.183 |
| MDL Number | MFCD00864296 |
| InChI Key | MNIPYSSQXLZQLJ-UHFFFAOYSA-N |
| Synonym | aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish |
| PubChem CID | 71771 |
| ChEBI | CHEBI:31159 |
| IUPAC Name | 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| SMILES | C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Chemical Identifiers
| 89796-99-6 | |
| 354.183 | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 71771 | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| C16H13Cl2NO4 | |
| MFCD00864296 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| CHEBI:31159 | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Specifications
| 152°C | |
| Yellow | |
| MFCD00864296 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl | |
| 1 g | |
| 71771 | |
| 354.18 | |
| Crystalline Powder |
| 89796-99-6 | |
| C16H13Cl2NO4 | |
| 2811 | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid | |
| 354.183 | |
| CHEBI:31159 | |
| ≥98.0% (HPLC,T) | |
| Aceclofenac |
Safety and Handling
RTECSNumber : CY1577900
TSCA : No