missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aceclofenac 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 152°C |
| Color | Yellow |
| Packaging | Glass Bottle |
| UN Number | 2811 |
| Quantity | 5 g |
| Formula Weight | 354.18 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Aceclofenac |
Safety and Handling
| RTECSNumber | CY1577900 |
| TSCA | No |