missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Accutase - Enzyme Cell Detachment Medium

Catalog No. 501129055
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129055 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501129055 Supplier Invitrogen™ Supplier No. 00455556

Invitrogen™ Accutase Enzyme Cell Detachment Medium performs exceptionally well in detaching cells for analysis of cell surface markers and virus growth assay.

  • Accutase performs exceptionally well in detaching cells for analysis of cell surface markers, virus growth assay, quiescence assays by serum starvation, transformation assays by oncogene transfection, neural crest cell migration assays, cell proliferation, cell haptotaxsis, tumor cell migration assays, routine cell passage, production scale-up (bioreactor), and flow cytometry.
  • Accutase has been shown effective on: fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, vascular smooth muscle cells, hepatocyte progenitors, primary chick embryo neuronal cells, bone marrow stem cells, adherent CHO and BHK cells, macrophages, 293 cells, L929 cells, immortalized mouse testicular germ cells, 3T3, Vero, COS, HeLa, NT2, MG63, M24 and A375 metastatic melanoma, gliomas U251 and D54, HT1080 fibrosarcoma cells, and Sf9 insect cells.

Specifications

Cell Type Mammalian Cells
Form Liquid
Product Type Cell Culture Dissociation Reagent
With Additives Phenol Red
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.