missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Accutase™ Cell Detachment Solution, Liquid

Catalog No. MT25058CV
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25058CV 500 mL
MT25058CI 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT25058CV Supplier Corning™ Supplier No. 25058CV
Only null left

Efficient, with gentle detachment of adherent cells within minutes

Accutase is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including our selection of Advanced Surface coated plasticware. Accutase is useful for cell detachment and for preparing single-cell suspensions from clumped cell populations for sub-culturing cells, analytical studies, and for accurate cell counting.

Accutase is formulated at a concentration that is ready to use, once thawed. (Note: Never thaw Accutase at 37°C.) A thawed bottle of Accutase can be removed from the refrigerator and immediately applied to cells. It should not be pre-warmed to 37°C. Accutase contains proteolytic activity that gently breaks down cellular adhesion molecules and enables cell detachment from the bottom of the flask.

Accutase cell detachment solution is:

  • Efficient, with gentle detachment of adherent cells within minutes
  • An alternative to Trypsin
  • Able to eliminate neutralization or wash steps
  • Accutase does not contain mammalian or bacterial derived products

Specifications

Content And Storage Store at -20°C.
Form Liquid
Sterility Sterile
Formulation 1 X Accutase enzymes prepared in Dulbeccos PBS (0.2g/L KCl, 0.2g/L KH2PO5, 8g/L NaCl, and 1.15g/L Na2HPO4) containing 0.5 mm EDTA·4Na and 3 mg/L Phenol Red.
Quantity 500 mL