Learn More
Fisherbrand™ accumet™ XL500 Benchtop Dual pH/ISE and Conductivity Meters
Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL500 dual pH/ISE/conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
Specifications
| Outputs | RS232, USB-A, Mini-B USB, RJ45, stirrer port (2) |
|---|---|
| Accuracy | Multiple |
| Accuracy (Conductivity) | ±1% FS |
| Accuracy (ISE) | 0.5% full scale (monovalent ion), 1% full scale (divalent ion) |
| Accuracy (mV) | ±0.2 mV |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
13636XL500
|
Fisherbrand™
13636XL500 |
Meter with Kit |
N/A
|
|
|||||
|
XL500CERT
|
Fisherbrand™
XL500CERT |
Meter with Kit, NIST Cert |
N/A
|
|
|||||
Description
Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL500 dual pH/ISE/conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.
- High resolution color display shows data clearly from wide viewing angles and distances
- Optimized touch-screen with glass/glass sensitivity provides great ease of use
- Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
- One to six point pH calibration with 0.001 pH resolution
- Automatic buffer recognition with six pH buffer sets or custom buffer option
- Two to six point ISE calibration with 0.001 ppm, mg/L or molar resolution
- ISE methods for known addition, known subtraction, analyte addition and analyte subtraction
- Conductivity, TDS (total dissolved solids), salinity and resistivity modes
- Automatic or manual temperature compensation, displayed in °C or °F
- Display up to three measurements simultaneously (two pH or ISE, one conductivity)
- Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
- RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
- RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
- Bidirectional communication for LIMS connectivity and 3rd party software
- Connectivity for peripherals such as keyboard, mouse or other USB device
- Multilanguage interface with English, German, Chinese, French, Italian and Spanish
- Two ports for optional stirring probe (13-620-BSP) with five speed settings
- Option to wall-mount meter to create additional bench space
NIST-Traceable Calibration Models (-CERT)
- Help meet ISO, FDA, USDA and EPA quality assurance guidelines
- Includes meter accuracy at five test points across pH range
- Includes meter accuracy at five test points across corresponding mV range
- Testing performed by accredited laboratory, includes NIST-Traceable certificate
Specifications
- pH Range: -2.000 to 20.000 pH
- pH Resolution: 0.1, 0.01, 0.001 pH
- pH Accuracy: ±0.002 pH
- ISE Range: 0.001 to 19999 ppm, mg/L or molar
- ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
- ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
- mV Range: ±2000.0mV
- mV Resolution: 0.1mV
- mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
- mV Offset Adjustment: Up to ±150mV
- Conductivity Range: 0.00μS to 500.0mS
- Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
- Conductivity Accuracy: ±1 % full scale
- TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
- TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
- TDS Accuracy: ±1 % full scale
- Salinity Range: 0 to 80.0 ppt
- Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
- Salinity Accuracy: ±1 % full scale
- Resistivity Range: 2.000 Ohm to 20.0 MOhm
- Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
- Resistivity Accuracy: ±1 % full scale
- Temperature Range: -5° to +105°C (23° to 221.0°F)
- Temperature Resolution: 0.1°C (0.1°F)
- Temperature Accuracy: ±0.2°C (±0.3°F)
- Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
- ISE Calibration Points: 2 to 6 points
- ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
- Memory: 2000 data sets per user ID with date/time stamp
- Self-Test: Yes
- Inputs: Two BNC, 2 mini-phone ATC, 8 Pin DIN (conductivity), ground
- Output: RJ45, RS232, 2 USB, two stirring probe
- Power Requirements: 100 to 240VAC, 50/60Hz
- Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
- Weight: 1.13kg (2.5 lb.)
Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage
Certifications
CE, TUV 3-1, FCC Class A
Warranty
3 years on meter
Specifications
| RS232, USB-A, Mini-B USB, RJ45, stirrer port (2) | |
| ±1% FS | |
| ±0.2 mV | |
| ± 1% full scale | |
| ± 1% full scale | |
| 2 to 6 | |
| 0.01 to 10 | |
| 3 to 86400 sec. | |
| Multiple | |
| 0 to 500 mS | |
| -2 to 20 | |
| 0 to 80 ppt | |
| 2/3/4 Digits | |
| 2 BNC, 2 ATC | |
| Multiple | |
| 0.1mV | |
| 0.01/0.1ppm, 0.001/0.01/0.1ppt | |
| ±0.3°C | |
| 3 Years | |
| 9.68 in. | |
| 17.3 cm | |
| 9.9 cm | |
| 1.13 kg | |
| 50/60Hz |
| Multiple | |
| 0.5% full scale (monovalent ion), 1% full scale (divalent ion) | |
| ±0.002 | |
| ± 1% full scale | |
| Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range | |
| Up to 6 preset or 5 custom | |
| NIST, ISO, FDA, USDA, EPA | |
| 2000 | |
| LCD | |
| ±2000 mV | |
| 3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW | |
| 0.050 ppm to 500 ppt (at TDS factor 1.00) | |
| 0.1/0.01/0.001 | |
| ±2000 mV | |
| 0.001/0.01/0.1mS | |
| 0.01/0.1W, 0.001/0.1kW, 0.01MW | |
| 0.01/0.1ppm, 0.001/0.01/0.1ppt | |
| 0.1°C | |
| 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.) | |
| 24.6 cm | |
| 3.89 in. | |
| 2.5 lb. | |
| 100/240 V |