missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ XL200 Benchtop pH/Conductivity Meters

Catalog No. XL200ACERT
Change view
Click to view available options
Product Type:
Meter with pH/Cond. Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
XL200ACERT Meter with pH/Cond. Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XL200ACERT Supplier Fisherbrand™ Supplier No. 5933109
Only null left

Includes

accumet XL200 pH/conductivity benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST traceability certificate

Provide advanced measurements with simple operation and offer a large, vivid color touch-screen display that allows for dynamic functionality. Fisherbrand accumet XL200 pH/Conductivity benchtop meters offer advanced features with simple operation to help ensure your measurements are reliable and your laboratory processes are efficient.

  • High resolution color display shows data clearly from wide viewing angles and distances
  • Optimized touch-screen with glass/glass sensitivity provides great ease of use
  • Onscreen info includes date and time, stability indicator, alarms and context-specific help screens
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with six pH buffer sets or custom buffer option
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Display up to three measurements simultaneously (two pH or ISE, one conductivity)
  • Up to 10 password protected user ID accounts, each retains unique calibration and instrument settings with memory to store 2000 data points
  • RS232 and USB interfaces for printer or computer interfacing and real-time onscreen graphing of measurements
  • RJ45 Ethernet/Internet port to connect Local Area Network (LAN) or Internet
  • Bidirectional communication for LIMS connectivity and 3rd party software
  • Connectivity for peripherals such as keyboard, mouse or other USB device
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Two ports for optional stirring probe (Cat. No. 13-620-BSP) with five speed settings
  • Option to wall-mount meter to create additional bench space

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • ISE Range: 0.001 to 19999 ppm, mg/L or molar
  • ISE Resolution: 0.001, 0.01, 0.1, 1 (automatic)
  • ISE Accuracy: 0.5 % full scale (monovalent ion); 1 % full scale (divalent ion)
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: -5° to +105°C (23° to 221.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.2°C (±0.3°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Memory: 2000 data sets per user ID with date/time stamp
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, 8 Pin DIN (conductivity), ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
  • Dimensions (L x W x H): 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
  • Weight: 1.13kg (2.5 lb.)

Applications
Pharmaceutical, biotech, research, QA/QC, industrial, food, beverage

Certifications

CE, TUV 3-1, FCC Class A

Warranty

3 years on meter

Specifications

Outputs RS232, USB-A, Mini-B USB, RJ45, stirrer port (2)
Accuracy Multiple
Accuracy (Conductivity) ±1% FS
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Accuracy (Resistivity) ± 1% full scale
Accuracy (Salinity) ± 1% full scale
Accuracy (TDS) ± 1% full scale
Calibration Points (Conductivity) Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (pH) Up to 6 preset or 5 custom
Cell Constant 0.01 to 10
Certifications/Compliance NIST, ISO, FDA, USDA, EPA
Data Management Details 3 to 86400 sec.
Data Points 2000
Detection Range Multiple
Display Type LCD
Range (Conductivity) 0 to 500 mS
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 3 W to 100 MW over 4 ranges: 0 to 20 kW, 20 to 200 kW, 200 kW to 2 MW, 2 to 100 MW
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.050 ppm to 500 ppt (at TDS factor 1.00)
Resolution (pH) 0.1/0.01/0.001
Includes Meter, electrode arm, RS-232 and USB cables, 110/220 V power supply, and NIST-traceable certificate
Inputs BNC, ATC
Model XL200 pH/Conductivity Meter w/ NIST-Traceable Certificate
Product Type Meter with pH/Cond. Kit
Resolution Multiple
Resolution (Conductivity) 0.001/0.01/0.1mS
Resolution (mV) 0.1mV
Resolution (Resistivity) 0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Temperature Accuracy ±0.2°C
Temperature Resolution 0.1°C
Warranty 3 Years
Dimensions (L x W x H) 24.6 x 17.3 x 9.9cm (9.7 x 6.8 x 3.9 in.)
Length (English) 9.68 in.
Length (Metric) 24.6 cm
Width (Metric) 17.3 cm
Height (English) 3.89 in.
Height (Metric) 9.9 cm
Weight (English) 2.5 lb.
Weight (Metric) 1.13 kg
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less