missing translation for 'onlineSavingsMsg'
Learn More
Learn More
accumet™ ROSS pH Electrode Storage Solution, Fisherbrand™
Specifications
Specifications
| Chemical Name or Material | pH Electrode Storage Solution |
| CAS | 7732-18-5, 7447-40-7, 877-24-7 |
| Certificate of Analysis | Yes |
| Description | ROSS pH Electrode Storage Solution, 1 pint (475 mL) Bottle |
| Concentration | 1X |
| For Use With (Application) | pH Measurement |
| For Use With (Equipment) | pH Electrode |
| Physical Form | Liquid |
| Quantity | 475 mL |
| Solution Type | Storage |
| Show More |