missing translation for 'onlineSavingsMsg'
Learn More

accumet™ pH Electrode Storage Solution, Fisherbrand™

Catalog No. 15200900
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15200900 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15200900 Supplier Thermo Scientific Supplier No. 910001FS
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material pH electrode solution
Color Colorless
Certificate of Analysis Yes
Description pH Electrode Storage Solution, 1 pint (475 mL) Bottle
For Use With (Application) Ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
For Use With (Equipment) pH Electrode
pH 6.6
Physical Form Liquid
Quantity 475 mL
Solution Type Storage
Packaging Bottle
Traceability to NIST Yes
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less