missing translation for 'onlineSavingsMsg'
Learn More

accumet™ pH Adjusting ISA for Ammonia Electrode, Fisherbrand™

Catalog No. 15200905
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15200905 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15200905 Supplier Thermo Scientific Supplier No. 951211FS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ion Selective Electrode Reagents
CAS 7732-18-5, 1310-73-2, 6381-92-6, 125-20-2, 67-56-1
Certificate of Analysis Yes
Description pH Adjusting ISA for Ammonia Electrode
Concentration 5 M
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
For Use With (Equipment) Electrode
Physical Form Liquid
Quantity 475 mL
Solution Type Ionic Strength Adjustor
Packaging Bottle
Traceability to NIST Yes
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less