missing translation for 'onlineSavingsMsg'
Learn More
Learn More
accumet™ pH 10.01 (Blue) Buffer, Fisherbrand™
Specifications
Specifications
| Chemical Name or Material | pH Buffer, bottles |
| Certificate of Analysis | Yes |
| Description | pH 10.01 (Blue) Buffer |
| For Use With (Application) | Accurate, reproducible pH calibrations |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 10.01 |
| Physical Form | Liquid |
| Quantity | 475 mL |
| Solution Type | Buffer |
| Packaging | Bottle |
| Show More |