missing translation for 'onlineSavingsMsg'
Learn More

accumet™ pH 1.68 Buffer, Fisherbrand™

Catalog No. 15200906
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15200906 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15200906 Supplier Thermo Scientific Supplier No. 910168FS
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material pH Buffer, bottles
Certificate of Analysis Yes
Description pH 1.68 Buffer - colorless
For Use With (Application) Accurate, reproducible pH calibrations
For Use With (Equipment) pH Meter and Electrode
pH 1.68
Physical Form Liquid
Quantity 475 mL
Solution Type Buffer
Packaging Bottle
Traceability to NIST Yes
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less