missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
Supplier: Fisherbrand™ AB315CERT
Description
- Large, backlit display simultaneous shows pH, mV and temperature readings
- One to five point pH calibration with automatic buffer recognition of four pH buffer sets
- One point relative mV calibration (RmV) for ORP measurements
- One point ATC temperature probe calibration
- Calibration due alarm, selectable 1 to 60 hr., or off
- Automatic or manual temperature compensation
- Ready icon indicates measurement stability
- Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
- Non-volatile meter memory preserves settings and data in event of power loss
- Meter-attached stand facilitates easy electrode movement into and out of samples
Chemical Identifiers
| ±0.2 or ±0.05%, Whichever is Greater | |
| ±0.3°C (±0.5°F) | |
| Port for Computer or Printer | |
| pH, Relative mV, Temperature | |
| 0.01, 0.1 | |
| ±2000 mV | |
| 1 to 5 (pH), 1 (Relative mV), 1 (Temperature) | |
| 23°F to 221°F | |
| 5 in. LCD | |
| Non-volatile | |
| Automatic, Manual | |
| IP54 | |
| 3 Years | |
| ±2000 mV | |
| Automatic, Manual | |
| 100/240 V |
| ±0.01 | |
| Printed or CSV Format | |
| pH with mV and Temperature, Relative mV with mV and Temperature | |
| -2 to 18 | |
| USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13 | |
| 1 mV, 0.1 RmV | |
| -5°C to +105°C | |
| 0.1°C (0.1°F) | |
| Yes | |
| 500 | |
| BNC, ATC | |
| Universal Power Adapter | |
| AB315 | |
| Indicator Icons | |
| 1 | |
| 50/60 Hz |
Specifications
| pH/mV Meter with Electrode Stand | |
| ±0.01 | |
| Port for Computer or Printer | |
| Electrode Stand, Power Adapter | |
| pH, Relative mV, Temperature | |
| 0.01, 0.1 | |
| ±2000 mV | |
| 1 to 5 (pH), 1 (Relative mV), 1 (Temperature) | |
| 23°F to 221°F | |
| 5 in. LCD | |
| Non-volatile | |
| Automatic, Manual | |
| CE, TUV 3-1, FCC Class A, With NIST-traceable Calibration Certificate | |
| Universal Power Adapter | |
| AB315 | |
| Indicator Icons | |
| 1 | |
| 100/240 V |
| ±0.3°C (±0.5°F) | |
| Printed or CSV Format | |
| ±0.2 or ±0.05%, Whichever is Greater | |
| pH with mV and Temperature, Relative mV with mV and Temperature | |
| -2 to 18 | |
| USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13 | |
| 1 mV, 0.1 RmV | |
| -5°C to +105°C | |
| 0.1°C (0.1°F) | |
| Yes | |
| 500 | |
| BNC, ATC | |
| IP54 | |
| 3 Years | |
| ±2000 mV | |
| Automatic, Manual | |
| pH/mV Meter with Probe, NIST | |
| 50/60 Hz |