missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter

Catalog No. p-7235485
Change view
Click to view available options
Type:
pH/mV Meter
pH/mV Meter with Probe
pH/mV Meter with Probe, NIST
pH/mV Meter, NIST
Probe Type:
Gel-filled pH/ATC Electrode
Refillable pH Electrode
Material:
Epoxy Body
Glass Body
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Probe Type Material
13636AB315 pH/mV Meter - -
13636AB315A pH/mV Meter with Probe Gel-filled pH/ATC Electrode Epoxy Body
13636AB315B pH/mV Meter with Probe Refillable pH Electrode Glass Body
AB315CERT pH/mV Meter with Probe, NIST - -
AB315ACERT pH/mV Meter, NIST Gel-filled pH/ATC Electrode Epoxy Body
AB315BCERT pH/mV Meter with Probe, NIST Refillable pH Electrode Glass Body
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13636AB315 Supplier Fisherbrand™ Supplier No. 13636AB315

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.

  • Large, backlit display simultaneous shows pH, mV and temperature readings
  • One to five point pH calibration with automatic buffer recognition of four pH buffer sets
  • One point relative mV calibration (RmV) for ORP measurements
  • One point ATC temperature probe calibration
  • Calibration due alarm, selectable 1 to 60 hr., or off
  • Automatic or manual temperature compensation
  • Ready icon indicates measurement stability
  • Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
  • Non-volatile meter memory preserves settings and data in event of power loss
  • Meter-attached stand facilitates easy electrode movement into and out of samples

Specifications

Description pH/mV Meter with Electrode Stand
Accuracy (pH) ±0.01
Data Management Details Printed or CSV Format
Accuracy (mV) ±0.2 or ±0.05%, Whichever is Greater
Outputs Port for Computer or Printer
Accuracy (Temperature) ±0.3°C (±0.5°F)
Includes Electrode Stand, Power Adapter
Measurement Modes pH with mV and Temperature, Relative mV with mV and Temperature
Calibration pH, Relative mV, Temperature
Range (pH) -2 to 18
Resolution (pH) 0.01, 0.1
Calibration Points (pH) USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
Range (mV) ±2000 mV
Resolution (mV) 1 mV, 0.1 RmV
Calibration Points 1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C (0.1°F)
Display Type 5 in. LCD
Backlight Option Yes
Memory Non-volatile
Data Points 500
Log Function Type Automatic, Manual
Inputs BNC, ATC
Certifications/Compliance CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate
IP Rating IP54
AC Adapter Universal Power Adapter
Warranty 3 Years
Model AB315
Range (Relative mV) ±2000 mV
Stability Indicator Icons
Temperature Compensation Automatic, Manual
Channels 1
Type pH/mV Meter
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less