missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter

Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.

Supplier:  Fisherbrand™ AB315ACERT

 View more versions of this product

Catalog No. AB315ACERT


Only null left

Description

Description

  • Large, backlit display simultaneous shows pH, mV and temperature readings
  • One to five point pH calibration with automatic buffer recognition of four pH buffer sets
  • One point relative mV calibration (RmV) for ORP measurements
  • One point ATC temperature probe calibration
  • Calibration due alarm, selectable 1 to 60 hr., or off
  • Automatic or manual temperature compensation
  • Ready icon indicates measurement stability
  • Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
  • Non-volatile meter memory preserves settings and data in event of power loss
  • Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications

Chemical Identifiers

±0.2 or ±0.05%, Whichever is Greater
±0.3°C (±0.5°F)
Port for Computer or Printer
pH, Relative mV, Temperature
0.01, 0.1
±2000 mV
1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
23°F to 221°F
5 in. LCD
Non-volatile
Automatic, Manual
IP54
3 Years
±2000 mV
Automatic, Manual
100/240 V
±0.01
Printed or CSV Format
pH with mV and Temperature, Relative mV with mV and Temperature
-2 to 18
USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
1 mV, 0.1 RmV
-5°C to +105°C
0.1°C (0.1°F)
Yes
500
BNC, ATC
Universal Power Adapter
AB315
Indicator Icons
1
50/60 Hz

Specifications

pH/mV Meter with pH/ATC Epoxy-body Double Junction Gel-filled Electrode
Port for Computer or Printer
±0.2 or ±0.05%, Whichever is Greater
pH/ATC Electrode, Electrode Stand, Power Adapter
pH, Relative mV, Temperature
0.01, 0.1
±2000 mV
1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
23°F to 221°F
5 in. LCD
Non-volatile
Automatic, Manual
CE, TUV 3-1, FCC Class A, With NIST-traceable Calibration Certificate
Universal Power Adapter
Epoxy Body
Gel-filled pH/ATC Electrode
Indicator Icons
1
100/240 V
Printed or CSV Format
±0.3°C (±0.5°F)
±0.01
pH with mV and Temperature, Relative mV with mV and Temperature
-2 to 18
USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
1 mV, 0.1 RmV
-5°C to +105°C
0.1°C (0.1°F)
Yes
500
BNC, ATC
IP54
3 Years
AB315
±2000 mV
Automatic, Manual
pH/mV Meter, NIST
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions