Learn More
Fisherbrand™ accumet™ AR425 pH/ion Meter with Stand
Designed to deliver precise and reliable electrochemistry testing, the accumet AR425 offers enhanced performance in a streamlined design. Ideal for laboratories requiring dependable measurement capabilities.
Supplier:  Fisherbrand™ 13636AR425
Catalog No. 13636AR425
Only null left
Description
Description
  • Large 7-in., high-resolution color display with capacitive touchscreen operable with gloves.
  • One to five point pH and Ion calibration with automatic buffer/standard recognition.
  • Streamlined, easy-to-operate user interface with step-by-step calibration guidance.
  • Stores 10 calibration logs per parameter with date and time.
  • Stores 5000 data log sets with date and time.
  • Outputs to a printer, computer, or USB flash drive for long-term data retention.
  • Inputs sample ID with optional USB barcode scanner, USB keyboard, or USB mouse.
  • Stirs samples with meter-controlled and meter-powered stirrer probe.
Specifications
Specifications
±0.2 mV or ±0.05% of the reading, whichever is greater
Yes
1
5000 with Date and Time Stamp
7-in. High-resolution Color Graphic LCD
pH, mV, ORP, Ion, Temperature
-2 to 20
-30.0 to +130.0°C
AR425
100/240 V
±0.002 pH
pH
CE, TUV 3-1, FCC Class A
1
IP54
-2000.0 to +2000.0 mV
0.1, 0.01, or 0.001 pH
Automatic Data Logging
Benchtop Multiparameter Meters
Product Suggestions
Product Suggestions