missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Accumax - Cell Aggregate Dissociation Medium

Catalog No. 509271
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
509271 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Contains azide.

The use of Accumax™ may effect the integrity of certain antigenic epitopes resulting in diminished antibody staining. This should be determined experimentally for individual antibody clones.

Catalog No. 509271 Supplier Invitrogen™ Supplier No. 00466656
  • ONLINE EXCLUSIVE PRICE
    $181.00
    $131.05 / Each

    Save $49.95

    28.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains azide.

The use of Accumax™ may effect the integrity of certain antigenic epitopes resulting in diminished antibody staining. This should be determined experimentally for individual antibody clones.

Dissociate cell clumps for cell counting, viral transfection assays, cell sorting, and flow cytometry as well as bioreactor scale-up.

Accumax has been shown effective in dissociating cell clumps in a variety of cell lines including hybridomas, CHO, BHK, 293, COS and Sf9 cells. The use of Accumax may effect the integrity of certain antigenic epitopes resulting in diminished antibody staining. This should be determined experimentally for individual antibody clones. Thaw Accumax at room temperature or 4°C.

Recommended Storage

Store at −20°C. Thawing during shipment will not affect performance.

Specifications

Cell Type Mammalian Cells
Form Liquid
Product Type Cell Culture Dissociation Reagent
Without Additives No Phenol Red
Quantity 100 mL

For Research Use Only. Not for use in diagnostic procedures.