Learn More
Accuform Signs U-Channel Posts
Mount signs to these rugged posts
Supplier: Accuform Signs HSP108
Description
- Posts can be set in ground
- Concrete or portable base
- 9.53mm (0.375 in.) holes on 25.4mm (1 in.) centers along the length
Order Info
3.1m (10 ft.) and larger posts are required to ship via truck. Shipping charges will be applied based upon weight and zone.
Specifications
| NCHRP-350 | |
| 1 Ea. | |
| 96 in. | |
| U-channel Sign post |
| 0.37 in. dia. Hole on 1 in. centers along the length | |
| To display signs either in temporary or permanent situations | |
| 2.4 m | |
| Green |