Learn More
Accuform Signs U-Channel Posts
Mount signs to these rugged posts
Supplier:  Accuform Signs HSP106
 View more versions of this product
Catalog No. 19804517
Only null left
Description
Description
  • Posts can be set in ground
  • Concrete or portable base
  • 9.53mm (0.375 in.) holes on 25.4mm (1 in.) centers along the length
Order Info
3.1m (10 ft.) and larger posts are required to ship via truck. Shipping charges will be applied based upon weight and zone.
Specifications
Chemical Identifiers
NCHRP-350
1 Ea.
U-channel Sign post
0.37 in. dia. Hole on 1 in. centers along the length
To display signs either in temporary or permanent situations
Specifications
NCHRP-350
1 Ea.
72 in.
U-channel Sign post
0.37 in. dia. Hole on 1 in. centers along the length
To display signs either in temporary or permanent situations
1.9 m
Green
Product Suggestions
Product Suggestions